Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120656.1 | 3 | 359 | |||
NR_120657.1 | 2 | 253 | ||||
NR_120658.1 | 2 | 249 | ||||
NR_120659.1 | 3 | 370 | ||||
GenBank mRNA | AK123067.1 | - | 3 | 359 | ||
DB454367.1 | - | 3 | 316 | |||
HY051619.1 | - | 2 | 199 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000411439.1 | - | 2 | 217 | Chr. 10 - 42674197 - 42691723 [-] on Build GRCh38 |