Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120656.1 | 1 | 47 | |||
NR_120657.1 | 1 | 47 | ||||
NR_120658.1 | 1 | 47 | ||||
NR_120659.1 | 1 | 47 | ||||
GenBank mRNA | AK123067.1 | - | 1 | 47 | ||
DB446959.1 | - | 1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000411439.1 | - | 11 | Chr. 10 - 42674197 - 42691723 [-] on Build GRCh38 |