Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034149.1 | 1 | 91 | |||
GenBank mRNA | AF258576.1 | - | 1 | 76 | ||
DA864592.1 | - | 1 | 91 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000443744.1 | - | 1 | 76 | Chr. 20 - 23125068 - 23132621 [-] on Build GRCh38 |