Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109772.1 | 4 | 884 | |||
GenBank mRNA | BC036431.2 | - | 4 | 884 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000437181.1 | - | 4 | 884 | Chr. 9 - 81689713 - 81776900 [+] on Build GRCh38 |