Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125940.1 | 4 | 463 | |||
NR_125941.1 | 3 | 415 | ||||
NR_125942.1 | 3 | 342 | ||||
GenBank mRNA | BU531772.1 | - | 3 | 345 |