Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125940.1 | 4 | 435 | |||
NR_125941.1 | 3 | 387 | ||||
NR_125942.1 | 3 | 314 | ||||
GenBank mRNA | BF175091.1 | - | 3 | 243 | ||
BU531772.1 | - | 3 | 317 |