Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125940.1 | 3, 4 | 368 | |||
NR_125941.1 | 2, 3 | 320 | ||||
NR_125942.1 | 2, 3 | 247 | ||||
GenBank mRNA | BF175091.1 | - | 2, 3 | 176 | ||
BU531772.1 | - | 2, 3 | 250 |