Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED14OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289773.1 | NP_001276702.1 | 2 | 493 | ||
GenBank mRNA | BC039399.1 | - | 2 | 489 |