Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015401.2 | 2 | 397 | |||
NR_072981.1 | 2 | 397 | ||||
NR_072982.1 | 2 | 263 | ||||
GenBank mRNA | AK092709.1 | - | 2 | 339 | ||
BC081571.1 | - | 2 | 275 | |||
DA066910.1 | - | 2 | 394 | |||
DA204840.1 | - | 2 | 234 | |||
DA212423.1 | - | 2 | 347 | |||
DA293391.1 | - | 2 | 232 | |||
DA406714.1 | - | 2 | 214 | |||
DA776150.1 | - | 2 | 397 |