Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108042.2 | 2 | 829 | |||
GenBank mRNA | BC042100.1 | - | 2 | 829 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457402.1 | - | 2 | 829 | Chr. 1 - 110487680 - 110490258 [-] on Build GRCh38 |