Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100507557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038244.1 | 4 | 3700 | |||
NR_038245.1 | 3 | 3594 | ||||
NR_038246.1 | 3 | 3126 | ||||
GenBank mRNA | AK124757.1 | - | 3 | 3124 | ||
AK311235.1 | - | 3 | 3558 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000606388.5 | - | 3 | 3558 | Chr. 6 - 145814912 - 145886585 [+] on Build GRCh38 |