Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038374.1 | 3 | 477 | |||
NR_038375.1 | 2 | 375 | ||||
NR_038376.1 | 2 | 254 | ||||
GenBank mRNA | BC028422.1 | - | 2 | 302 | ||
DB072413.1 | - | 2 | 255 | |||
DB091920.1 | - | 3 | 400 | |||
DB099527.1 | - | 2 | 211 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000443246.2 | - | 2 | 331 | Chr. 10 - 28789188 - 28796006 [-] on Build GRCh38 |