Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOA1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126362.1 | 2 | 199 | |||
GenBank mRNA | DA327409.1 | - | 2 | 199 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF960114.1 | - | 1 | 41 | Chr. 11 - 116855329 - 116855789 [+] on Build GRCh38 |