Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110919.1 | 2 | 160 | |||
GenBank mRNA | BU598223.1 | - | 2 | 162 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000424075.2 | - | 2 | 160 | Chr. 12 - 132887842 - 132888583 [+] on Build GRCh38 |