Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110267.1 | 3 | 1614 | |||
GenBank mRNA | AK096499.1 | - | 2 | 1326 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000456248.1 | - | 2 | 1331 | Chr. 2 - 198299363 - 198374549 [-] on Build GRCh38 |