Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ445850.1 | - | 1 | 481 |
Gene Symbol: PRKG1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038277.1 | 6 | 1215 | |||
GenBank mRNA | BC039504.2 | - | 1 | 29 | ||
BC050034.1 | - | 6 | 1217 |