Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034135.1 | 9 | 1216 | |||
GenBank mRNA | BC043563.1 | - | 8 | 976 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000430192.5 | - | 8 | 976 | Chr. 2 - 8007771 - 8324630 [-] on Build GRCh38 |