Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120632.1 | 4 | 1628 | |||
GenBank mRNA | AK055803.1 | - | 4 | 1629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455199.1 | - | 4 | 1628 | Chr. 10 - 4051726 - 4089013 [+] on Build GRCh38 |