Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002733.2 | 3 | 396 | |||
NR_045121.1 | 2 | 252 | ||||
NR_110533.1 | 3 | 396 | ||||
GenBank mRNA | BF313645.1 | - | 3 | 394 | ||
BG699848.1 | - | 2 | 252 | |||
X91348.1 | - | LIT1227 DGCR5 |
3 | 368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF317417.1 | - | 3 | 393 | Chr. 22 - 18970507 - 18990915 [+] on Build GRCh38 |