Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110285.1 | 4 | 1120 | |||
GenBank mRNA | AK123402.1 | - | 4 | 1120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000450840.1 | - | 4 | 1120 | Chr. 2 - 129923177 - 129946703 [+] on Build GRCh38 |