Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRY-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103839.1 | 2 | 243 | |||
GenBank mRNA | BC035084.1 | - | 3 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000428419.1 | - | 2 | 243 | Chr. 13 - 32025314 - 32031639 [-] on Build GRCh38 |