Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110258.2 | 5 | 1788 | |||
GenBank mRNA | BC042876.1 | - | 5 | 1788 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446838.2 | - | 5 | 1788 | Chr. 2 - 162768936 - 162797972 [+] on Build GRCh38 |