Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110086.1 | 5 | 597 | |||
NR_110087.1 | 3 | 219 | ||||
NR_110088.1 | 2 | 174 | ||||
GenBank mRNA | BG213706.1 | - | 1 | 95 | ||
BG256720.1 | - | 5 | 563 | |||
BM152701.1 | - | 3 | 214 | |||
CB217567.1 | - | 2 | 174 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435695.1 | - | 2 | 174 | Chr. 7 - 92836483 - 92917187 [+] on Build GRCh38 |