Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110797.1 | 4 | 899 | |||
GenBank mRNA | BC034007.1 | - | 4 | 904 | ||
BC045735.1 | - | 4 | 899 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431347.1 | - | 4 | 899 | Chr. 1 - 220829255 - 220832429 [+] on Build GRCh38 |