Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMMSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110000.1 | 4 | 1226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000483525.2 | - | 4 | 1237 | Chr. 3 - 69999744 - 70015318 [+] on Build GRCh38 |