Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110017.1 | 4 | 1650 | |||
GenBank mRNA | BC033333.1 | - | 4 | 1652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000481624.1 | - | 4 | 1650 | Chr. 3 - 112736447 - 112749319 [-] on Build GRCh38 |