Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC148638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK058184.1 | - | 3 | 399 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441360.1 | - | 3 | 399 | Chr. 1 - 234660271 - 234667104 [+] on Build GRCh38 |