Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104146.1 | 8 | 5448 | |||
GenBank mRNA | BX641067.1 | - | 1 | 2178 | ||
CR749396.1 | - | 1 | 1368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000482787.7 | - | 8 | 5469 | Chr. 3 - 181952343 - 182010678 [+] on Build GRCh38 |