Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104147.1 | 1 | 787 | |||
GenBank mRNA | AK097794.1 | - | 1 | 787 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000475981.5 | - | 1 | 787 | Chr. 3 - 158545220 - 158571066 [-] on Build GRCh38 |