Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105377105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135534.1 | 2 | 1857 | |||
GenBank mRNA | BC041347.2 | - | 2 | 1840 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000488720.1 | - | 2 | 1858 | Chr. 3 - 58162547 - 58170636 [-] on Build GRCh38 |