Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033843.1 | 7 | 995 | |||
GenBank mRNA | AK093355.1 | - | 7 | 988 | ||
BC043538.1 | - | 6 | 664 | |||
BI828519.1 | - | 6 | 664 |