Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK15-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132377.1 | 2 | 301 | |||
GenBank mRNA | BF195580.1 | - | 2 | 95 | ||
DB261503.1 | - | 2 | 300 |
Gene Symbol: LINC01260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090842.1 | - | 1 | 71 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000445420.5 | - | 2 | 69 | Chr. 20 - 44695202 - 44745995 [-] on Build GRCh38 |