Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080475.2 | NP_001073944.1 | 8 | 8706 | ||
XM_005246545.3 | XP_005246602.1 | 7 | 8548 | |||
XM_011511160.2 | XP_011509462.1 | 9 | 9041 | |||
XM_017004072.1 | XP_016859561.1 | 7 | 8891 | |||
XM_017004073.1 | XP_016859562.1 | 7 | 7770 | |||
GenBank mRNA | AF462444.1 | - | 1 | 386 | ||
BC042862.1 | - | 1 | 381 | |||
BX538248.1 | - | 1 | 742 |