Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135072.1 | 2 | 1674 | |||
GenBank mRNA | AK093020.1 | - | 2 | 1674 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501648.2 | - | 2 | 1674 | Chr. 11 - 128681240 - 128686922 [-] on Build GRCh38 |