Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105375650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132986.1 | 2 | 2049 | |||
NR_132987.1 | 2 | 1999 |
Gene Symbol: LOC286149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093190.1 | - | 2 | 1999 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501164.2 | - | 2 | 1999 | Chr. 8 - 95066808 - 95072920 [-] on Build GRCh38 |