Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024383.1 | 4 | 683 | |||
NR_024384.1 | 3 | 885 | ||||
GenBank mRNA | AK091356.1 | - | 4 | 1498 | ||
AK308694.1 | - | 3 | 884 | |||
AK309800.1 | - | 3 | 880 | |||
AK311230.1 | - | 3 | 875 | |||
BC036480.2 | - | 4 | 705 | |||
BC047100.1 | - | 3 | 693 | |||
BC048008.1 | - | 3 | 693 |