Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120563.1 | 3 | 1516 | |||
GenBank mRNA | AL833634.1 | - | 3 | 1469 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500537.2 | - | 3 | 1516 | Chr. 11 - 112959279 - 112963460 [-] on Build GRCh38 |