Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103771.1 | 3 | 3342 | |||
GenBank mRNA | AK126802.1 | - | 3 | 3342 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000503989.1 | - | 3 | 3342 | Chr. 5 - 6686325 - 6707711 [-] on Build GRCh38 |