Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102467224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104997.1 | 3 | 679 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000509367.1 | - | 2 | 634 | Chr. 5 - 118187467 - 118266035 [+] on Build GRCh38 |