Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUNAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038861.1 | 2 | 452 | |||
NR_132398.1 | 3 | 585 | ||||
NR_132399.1 | 2 | 194 | ||||
GenBank mRNA | AK023027.1 | - | 2 | 195 | ||
AK091547.1 | - | 2 | 408 | |||
BC016484.1 | - | 2 | 253 | |||
DA412350.1 | - | 2 | 452 |