Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103753.1 | 3 | 727 | |||
NR_103754.1 | 4 | 712 | ||||
NR_103755.1 | 3 | 649 | ||||
NR_103756.1 | 2 | 578 | ||||
GenBank mRNA | AK092078.1 | - | 2 | 869 | ||
BG777293.1 | - | 2 | 532 | |||
CN270888.1 | - | 4 | 555 | |||
DA458115.1 | - | 3 | 546 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000510145.1 | - | 2 | 673 | Chr. 5 - 102609156 - 102671559 [-] on Build GRCh38 |