Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103753.1 | 3 | 759 | |||
NR_103754.1 | 4 | 744 | ||||
NR_103755.1 | 3 | 681 | ||||
NR_103756.1 | 2 | 610 | ||||
GenBank mRNA | AK092078.1 | - | 2 | 901 | ||
BG777293.1 | - | 2 | 564 | |||
CN270888.1 | - | 4 | 587 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000510145.1 | - | 2 | 705 | Chr. 5 - 102609156 - 102671559 [-] on Build GRCh38 |