Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125040.1 | - | 3 | 1194 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000513405.1 | - | 2 | 1194 | Chr. 11 - 134735596 - 134763810 [+] on Build GRCh38 |