Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC286114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033894.1 | 1 | 589 | |||
GenBank mRNA | AK058067.1 | - | 1 | 589 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000522604.1 | - | 1 | 589 | Chr. 8 - 20973986 - 20995119 [+] on Build GRCh38 |