Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105375875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093370.1 | - | 1 | 96 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000521221.5 | - | 1 | 96 | Chr. 8 - 63384839 - 63417930 [-] on Build GRCh38 |