Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027012.1 | 3 | 185 | |||
NR_027013.1 | 3 | 185 | ||||
GenBank mRNA | AK097475.1 | - | 3 | 185 | ||
BC041354.1 | - | 3 | 197 |