Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002323.2 | 2 | 2690 | |||
NR_110492.1 | 3 | 2746 | ||||
NR_110493.1 | 2 | 2690 | ||||
GenBank mRNA | AK000625.1 | - | 2 | 689 | ||
AK000749.1 | - | 3 | 590 | |||
AK056480.1 | - | 2 | 1224 | |||
AK057695.1 | - | 1 | 714 | |||
BC037986.1 | - | 2 | 847 | |||
CR456447.1 | - | 2 | 382 |