Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510676.2 | XP_011508978.1 | 23 | 4760 |
Gene Symbol: OSGEPL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102429.1 | 1 | 77 | |||
GenBank mRNA | BX474356.1 | - | 1 | 154 | ||
DB142079.1 | - | 1 | 36 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000520651.1 | - | 1 | 153 | Chr. 2 - 189762704 - 189765556 [+] on Build GRCh38 |