Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038875.2 | 2 | 2100 | |||
GenBank mRNA | BC037345.2 | - | 2 | 2100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000519550.1 | - | 2 | 2100 | Chr. 8 - 63769430 - 63785501 [+] on Build GRCh38 |