Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100288181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038925.1 | 3 | 4724 | |||
GenBank mRNA | CR627188.1 | - | 3 | 4726 |
Gene Symbol: LY6K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB528496.1 | - | 1 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000520572.1 | - | 3 | 4724 | Chr. 8 - 142702252 - 142726973 [-] on Build GRCh38 |